Ethyl 4,6-dichloro-2-(methylthio)quinoline-3-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR8002-1G OR8002-500MG
APOSOR8002-1GEA 785 DKK
APOSOR8002-1G APOSOR8002-500MG
Ethyl 4,6-dichloro-2-(methylthio)quinoline-3-carboxylate
Ethyl 4,6-dichloro-2-(methylthio)quinoline-3-carboxylate
Formel: C₁₃H₁₁Cl₂NO₂S
Smeltepunkt: 133…135 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00276616
CAS-nummer: 227958-96-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR