3-Phenylcyclobutan-1-one
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Phenylcyclobutanone
OR7098-1G
OR7098-5G
APOSOR7098-1GEA
578
DKK
APOSOR7098-1G
APOSOR7098-5G
3-Phenylcyclobutan-1-one
3-Phenylcyclobutan-1-one
Formel:
C₁₀H₁₀O Molekylvægt: 146.186 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04038993 CAS-nummer: 52784-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...