1-Methyl-3-(3-methylcyclohex-1-yl)benzene
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR70207-1G
OR70207-250MG
OR70207-5G
APOSOR70207-1GEA
2650
DKK
APOSOR70207-1G
APOSOR70207-250MG
APOSOR70207-5G
1-Methyl-3-(3-methylcyclohex-1-yl)benzene
Formel:
C₁₄H₂₀ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD28122918 CAS-nummer: 154427-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...