4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbonitrile 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR70055-25G
OR70055-5G
OR70055-1G
APOSOR70055-25GEA
1170
DKK
APOSOR70055-25G
APOSOR70055-5G
APOSOR70055-1G
4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbonitrile 98%
4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbonitrile
Formel:
C₆H₄ClN₃S Smeltepunkt: 67…68 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00507261 CAS-nummer: 33089-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...