Methyl 5-methylhex-2-enoate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR6768-1G OR6768-25G OR6768-5G
APOSOR6768-1GEA 284 DKK
APOSOR6768-1G APOSOR6768-25G APOSOR6768-5G
Methyl 5-methylhex-2-enoate 97%
Methyl 5-methylhex-2-enoate
Formel: C₈H₁₄O₂
Kogepunkt: 95…100 °C (26 mm Hg)
Flammepunkt: 57 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00137613
CAS-nummer: 68797-67-1
UN nummer: 3272
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR