3-(Chloromethyl)thiophene
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(Thien-3-yl)methyl chloride
OR6634-1G
OR6634-5G
APOSOR6634-1GEA
2770
DKK
APOSOR6634-1G
APOSOR6634-5G
3-(Chloromethyl)thiophene
3-(Chloromethyl)thiophene
Formel:
C₅H₅ClS Kogepunkt: 74…75 °C (14 mm Hg) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD08445657 CAS-nummer: 2746-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...