2-Chloro-6-methoxyisonicotinaldehyde

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloro-4-formyl-6-methoxypyridine, 2-Chloro-6-methoxypyridine-4-carboxaldehyde, 2-Chloro-6-methoxy-pyridine-4-carbaldehyde

OR61392-1G OR61392-500MG
APOSOR61392-1GEA 6710 DKK
APOSOR61392-1G APOSOR61392-500MG
2-Chloro-6-methoxyisonicotinaldehyde
2-Chloro-6-methoxyisonicotinaldehyde
Formel: C₇H₆ClNO₂
Molekylvægt: 171.581 g/mol
Smeltepunkt: 69…71 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06410665
CAS-nummer: 329794-31-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR