Methyl 2,4,5-trichlorobenzoate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR61081-1G OR61081-250MG OR61081-5G
APOSOR61081-1GEA 2100 DKK
APOSOR61081-1G APOSOR61081-250MG APOSOR61081-5G
Methyl 2,4,5-trichlorobenzoate
Methyl 2,4,5-trichlorobenzoate
Formel: C₈H₅Cl₃O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02149449
CAS-nummer: 86569-81-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR