(3-Bromophenyl)methanesulphonyl chloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR6077-1G OR6077-25G OR6077-5G
APOSOR6077-1GEA 240 DKK
APOSOR6077-1G APOSOR6077-25G APOSOR6077-5G
(3-Bromophenyl)methanesulphonyl chloride 97%
(3-Bromophenyl)methanesulphonyl chloride
Formel: C₇H₆BrClO₂S
Smeltepunkt: 88…89 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06798043
CAS-nummer: 58236-74-1
UN nummer: 3261
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR