6-Chloro-1H-imidazo[4,5-c]pyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Chloro-6-azabenzimidazole, 6-Chloro-3H-imidazo[4,5-c]pyridine

OR60107-1G OR60107-250MG
APOSOR60107-1GEA 2170 DKK
APOSOR60107-1G APOSOR60107-250MG
6-Chloro-1H-imidazo[4,5-c]pyridine
6-Chloro-1H-imidazo[4,5-c]pyridine
Formel: C₆H₄ClN₃
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12022343
CAS-nummer: 2589-11-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR