1,3-Oxazole-4-carbonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR60008-1G OR60008-250MG
APOSOR60008-1GEA 495 DKK
APOSOR60008-1G APOSOR60008-250MG
1,3-Oxazole-4-carbonitrile
1,3-Oxazole-4-carbonitrile
Formel: C₄H₂N₂O
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08059302
CAS-nummer: 55242-84-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR