6-Chlorobenzofuran

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR55738-1G OR55738-250MG
APOSOR55738-1GEA 4080 DKK
APOSOR55738-1G APOSOR55738-250MG
6-Chlorobenzofuran
6-Chlorobenzofuran
Formel: C₈H₅ClO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18825837
CAS-nummer: 151619-12-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR