2,7-Dibromo-9-(hydroxymethyl)fluorene 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2,7-Dibromo-9H-fluoren-9-yl)methanol

OR55504-1G OR55504-5G
APOSOR55504-1GEA 1370 DKK
APOSOR55504-1G APOSOR55504-5G
2,7-Dibromo-9-(hydroxymethyl)fluorene 95%
2,7-Dibromo-9-(hydroxymethyl)fluorene
Formel: C₁₄H₁₀Br₂O
Molekylvægt: 354.037 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18384775
CAS-nummer: 74316-23-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR