8-Iodo-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
OR55435-1G
OR55435-250MG
APOSOR55435-1GEA
7580
DKK
APOSOR55435-1G
APOSOR55435-250MG
8-Iodo-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%
8-Iodo-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide
Formel:
C₁₀H₁₀INOS Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD31629913 CAS-nummer: 2169310-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...