1-Bromodibenzo[b,d]furan-4-ol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR55146-1G OR55146-250MG
APOSOR55146-1GEA 4310 DKK
APOSOR55146-1G APOSOR55146-250MG
1-Bromodibenzo[b,d]furan-4-ol
1-Bromodibenzo[b,d]furan-4-ol
Formel: C₁₂H₇BrO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD30746962
CAS-nummer: 873974-43-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR