trans-4-Methoxycyclohexan-1-amine 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
trans-4-Methoxycyclohexanamine
OR54676-1G
OR54676-250MG
APOSOR54676-1GEA
709
DKK
APOSOR54676-1G
APOSOR54676-250MG
trans-4-Methoxycyclohexan-1-amine 95%
trans-4-Methoxycyclohexan-1-amine
Formel:
C₇H₁₅NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD20278175 CAS-nummer: 121588-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...