4-Bromo-3-formylbenzonitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR52617-1G
OR52617-250MG
APOSOR52617-1GEA
1220
DKK
APOSOR52617-1G
APOSOR52617-250MG
4-Bromo-3-formylbenzonitrile
4-Bromo-3-formylbenzonitrile
Formel:
C₈H₄BrNO Molekylvægt: 210.027 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11858309 CAS-nummer: 89003-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...