1-(4-Bromo-2-methylphenyl)ethanone
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR52566-25G
OR52566-5G
APOSOR52566-25GEA
1980
DKK
APOSOR52566-25G
APOSOR52566-5G
1-(4-Bromo-2-methylphenyl)ethanone
1-(4-Bromo-2-methylphenyl)ethanone
Formel:
C₉H₉BrO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11112126 CAS-nummer: 65095-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...