4-Phenylpyrimidine-2-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR52152-1G
OR52152-250MG
APOSOR52152-1GEA
2640
DKK
APOSOR52152-1G
APOSOR52152-250MG
4-Phenylpyrimidine-2-carboxylic acid
4-Phenylpyrimidine-2-carboxylic acid
Formel:
C₁₁H₈N₂O₂ Molekylvægt: 200.193 g/mol Smeltepunkt: 154…157 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD16988171 CAS-nummer: 74647-39-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...