3-Bromo-5-cyanobenzoic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR480841-1G OR480841-5G
APOSOR480841-1GEA 1870 DKK
APOSOR480841-1G APOSOR480841-5G
3-Bromo-5-cyanobenzoic acid 97%
3-Bromo-5-cyanobenzoic acid
Formel: C₈H₄BrNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09261037
CAS-nummer: 453566-14-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR