2-Amino-2-(4-methylsulphanylphenyl)acetic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR480448-1G OR480448-2G OR480448-5G
APOSOR480448-1GEA 1290 DKK
APOSOR480448-1G APOSOR480448-2G APOSOR480448-5G
2-Amino-2-(4-methylsulphanylphenyl)acetic acid
2-Amino-2-(4-methylsulphanylphenyl)acetic acid
Formel: C₉H₁₁NO₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02662405
CAS-nummer: 7292-80-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR