(E)-4-(3-Butoxyacryloyl)benzonitrile
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR480287-1G
OR480287-250MG
OR480287-500MG
APOSOR480287-1GEA
6470
DKK
APOSOR480287-1G
APOSOR480287-250MG
APOSOR480287-500MG
(E)-4-(3-Butoxyacryloyl)benzonitrile
(E)-4-(3-Butoxyacryloyl)benzonitrile
Formel:
C₁₄H₁₅NO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD22683247 CAS-nummer: 1379801-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...