2-Bromo-5-isopropylthiazole 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR480272-1G OR480272-250MG OR480272-500MG
APOSOR480272-1GEA 691 DKK
APOSOR480272-1G APOSOR480272-250MG APOSOR480272-500MG
2-Bromo-5-isopropylthiazole 95%
2-Bromo-5-isopropylthiazole
Formel: C₆H₈BrNS
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12033480
CAS-nummer: 1159817-16-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR