5-Phenylpyridine-2-carbaldehyde 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR480265-1G
OR480265-250MG
OR480265-5G
APOSOR480265-1GEA
2880
DKK
APOSOR480265-1G
APOSOR480265-250MG
APOSOR480265-5G
5-Phenylpyridine-2-carbaldehyde 95%
5-Phenylpyridine-2-carbaldehyde
Formel:
C₁₂H₉NO Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD10700050 CAS-nummer: 780800-85-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...