4-Amino-2-cyanobenzoic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR47860-1G OR47860-5G
APOSOR47860-1GEA 1750 DKK
APOSOR47860-1G APOSOR47860-5G
4-Amino-2-cyanobenzoic acid 95%
4-Amino-2-cyanobenzoic acid
Formel: C₈H₆N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD24039083
CAS-nummer: 1369820-01-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR