Methyl 3-(4,6-Dichloro-5-pyrimidyl)butyrate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR471458-1G
OR471458-250MG
OR471458-5G
APOSOR471458-1GEA
2980
DKK
APOSOR471458-1G
APOSOR471458-250MG
APOSOR471458-5G
Methyl 3-(4,6-Dichloro-5-pyrimidyl)butyrate
Methyl 3-(4,6-Dichloro-5-pyrimidyl)butyrate
Formel:
C₉H₁₀Cl₂N₂O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD27943097 CAS-nummer: 1295516-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...