4-(Bromomethyl)quinoline
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR471097-1G
OR471097-250MG
APOSOR471097-1GEA
2670
DKK
APOSOR471097-1G
APOSOR471097-250MG
4-(Bromomethyl)quinoline
4-(Bromomethyl)quinoline
Formel:
C₁₀H₈BrN Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09953523 CAS-nummer: 5632-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...