4,6-Dihydroxynicotinamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR470170-1G OR470170-5G
APOSOR470170-1GEA 1490 DKK
APOSOR470170-1G APOSOR470170-5G
4,6-Dihydroxynicotinamide
4,6-Dihydroxynicotinamide
Formel: C₆H₆N₂O₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18651680
CAS-nummer: 5466-41-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR