3-[4-(Methylthio)phenoxy]propionic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR470107-1G
OR470107-5G
APOSOR470107-1GEA
1920
DKK
APOSOR470107-1G
APOSOR470107-5G
3-[4-(Methylthio)phenoxy]propionic acid
3-[4-(Methylthio)phenoxy]propionic acid
Formel:
C₁₀H₁₂O₃S Molekylvægt: 212.265 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD16883062 CAS-nummer: 18333-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...