Methyl N-Cbz-homomorpholine-2-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR470090-1G OR470090-5G
APOSOR470090-1GEA 2980 DKK
APOSOR470090-1G APOSOR470090-5G
Methyl N-Cbz-homomorpholine-2-carboxylate
Methyl N-Cbz-homomorpholine-2-carboxylate
Formel: C₁₅H₁₉NO₅
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16495878
CAS-nummer: 1226776-84-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR