4-Chloro-2-morpholinobenzoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR470069-1G OR470069-5G
APOSOR470069-1GEA 1920 DKK
APOSOR470069-1G APOSOR470069-5G
4-Chloro-2-morpholinobenzoic acid
4-Chloro-2-morpholinobenzoic acid
Formel: C₁₁H₁₂ClNO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13191619
CAS-nummer: 1197193-12-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR