6-Ethyloctahydropyrrolo[3,4-b]pyridine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR470026-1G
OR470026-5G
APOSOR470026-1GEA
4520
DKK
APOSOR470026-1G
APOSOR470026-5G
6-Ethyloctahydropyrrolo[3,4-b]pyridine
6-Ethyloctahydropyrrolo[3,4-b]pyridine
Formel:
C₉H₁₈N₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD12198849 CAS-nummer: 1141669-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...