3-(tert-Butoxy)benzeneboronic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR46618-1G
OR46618-250MG
OR46618-5G
APOSOR46618-1GEA
1910
DKK
APOSOR46618-1G
APOSOR46618-250MG
APOSOR46618-5G
3-(tert-Butoxy)benzeneboronic acid
3-(tert-Butoxy)benzeneboronic acid
Formel:
C₁₀H₁₅BO₃ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06801745 CAS-nummer: 312931-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...