2-Bromo-6-isopropylphenol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 6-Bromo-2-isopropylphenol

OR46615-1G OR46615-250MG OR46615-5G
APOSOR46615-1GEA 5400 DKK
APOSOR46615-1G APOSOR46615-250MG APOSOR46615-5G
2-Bromo-6-isopropylphenol
2-Bromo-6-isopropylphenol
Formel: C₉H₁₁BrO
Molekylvægt: 215.087 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16997221
CAS-nummer: 129976-32-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR