2-Bromo-3-iodoaniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR46249-1G OR46249-250MG
APOSOR46249-1GEA 4350 DKK
APOSOR46249-1G APOSOR46249-250MG
2-Bromo-3-iodoaniline
2-Bromo-3-iodoaniline
Formel: C₆H₅BrIN
Molekylvægt: 297.919 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18391614
CAS-nummer: 111721-76-7
UN nummer: 2811
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR