1-Boc-3-Fmoc-amino-3-piperidinecarboxylic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR452036-1G OR452036-250MG OR452036-500MG
APOSOR452036-1GEA 7850 DKK
APOSOR452036-1G APOSOR452036-250MG APOSOR452036-500MG
1-Boc-3-Fmoc-amino-3-piperidinecarboxylic acid 97%
1-Boc-3-Fmoc-amino-3-piperidinecarboxylic acid
Formel: C₂₆H₃₀N₂O₆
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02683125
CAS-nummer: 368866-21-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR