(R)-2-Amino-5-methylhex-4-enoic acid 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR452021-1G OR452021-500MG
APOSOR452021-1GEA 4910 DKK
APOSOR452021-1G APOSOR452021-500MG
(R)-2-Amino-5-methylhex-4-enoic acid 97%
(R)-2-Amino-5-methylhex-4-enoic acid
Formel: C₇H₁₃NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10565737
CAS-nummer: 81177-54-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR