Sodium oxazole-2-carboxylate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR45140-250MG OR45140-1G OR45140-5G
APOSOR45140-250MGEA 559 DKK
APOSOR45140-250MG APOSOR45140-1G APOSOR45140-5G
Sodium oxazole-2-carboxylate 97%
Sodium oxazole-2-carboxylate
Formel: C₄H₂NO₃·Na
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17926331
CAS-nummer: 1255098-88-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR