1-Bromo-2-(3-bromoprop-1-ynyl)benzene 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-Bromo-2-(3-bromoprop-1-yn-1-yl)benzene

OR42052-1G OR42052-250MG OR42052-5G
APOSOR42052-1GEA 2970 DKK
APOSOR42052-1G APOSOR42052-250MG APOSOR42052-5G
1-Bromo-2-(3-bromoprop-1-ynyl)benzene 96%
1-Bromo-2-(3-bromoprop-1-ynyl)benzene
Formel: C₉H₆Br₂
Molekylvægt: 273.952 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD22482445
CAS-nummer: 1228107-22-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR