1-[(1-Methylpiperidin-4-yl)methyl]piperazine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Methyl-4-[(piperazin-1-yl)methyl]piperidine, 1-((1-Methylpiperidin-4-yl)methyl)piperazine
OR4164-1G
OR4164-250MG
OR4164-5G
APOSOR4164-1GEA
665
DKK
APOSOR4164-1G
APOSOR4164-250MG
APOSOR4164-5G
1-[(1-Methylpiperidin-4-yl)methyl]piperazine
1-[(1-Methylpiperidin-4-yl)methyl]piperazine
Formel:
C₁₁H₂₃N₃ Kogepunkt: 77…78 °C (0,38 mm Hg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02093539 CAS-nummer: 496808-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...