3-Bromo-5-iodobenzyl bromide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: α,3-Dibromo-5-iodotoluene, 1-Bromo-3-(bromomethyl)-5-iodobenzene

OR40662-1G OR40662-250MG
APOSOR40662-1GEA 1310 DKK
APOSOR40662-1G APOSOR40662-250MG
3-Bromo-5-iodobenzyl bromide
3-Bromo-5-iodobenzyl bromide
Formel: C₇H₅Br₂I
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18391671
CAS-nummer: 1261553-05-2
UN nummer: 3261
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR