8-Bromo-3-chloroisoquinoline
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
8-Bromo-3-chloro-2-azanaphthalene
OR40226-250MG
OR40226-1G
APOSOR40226-250MGEA
1090
DKK
APOSOR40226-250MG
APOSOR40226-1G
8-Bromo-3-chloroisoquinoline
8-Bromo-3-chloroisoquinoline
Formel:
C₉H₅BrClN Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11044698 CAS-nummer: 1029720-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...