5-Bromo-2,4-dichloroquinazoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR400855-1G OR400855-250MG
APOSOR400855-1GEA 3000 DKK
APOSOR400855-1G APOSOR400855-250MG
5-Bromo-2,4-dichloroquinazoline
5-Bromo-2,4-dichloroquinazoline
Formel: C₈H₃BrCl₂N₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09954889
CAS-nummer: 134517-54-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR