2-Bromo-4-cyanobenzoic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR400423-1G
OR400423-250MG
OR400423-5G
APOSOR400423-1GEA
1640
DKK
APOSOR400423-1G
APOSOR400423-250MG
APOSOR400423-5G
2-Bromo-4-cyanobenzoic acid 95%
2-Bromo-4-cyanobenzoic acid
Formel:
C₈H₄BrNO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11847003 CAS-nummer: 956218-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...