2-Bromo-6-iodoaniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR400089-5G OR400089-1G OR400089-250MG
APOSOR400089-5GEA 1590 DKK
APOSOR400089-5G APOSOR400089-1G APOSOR400089-250MG
2-Bromo-6-iodoaniline
2-Bromo-6-iodoaniline
Formel: C₆H₅BrIN
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18391615
CAS-nummer: 84483-27-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR