3-Methyl-isoxazole-5-carbaldehyde

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR346321-1G OR346321-250MG
APOSOR346321-1GEA 2730 DKK
APOSOR346321-1G APOSOR346321-250MG
3-Methyl-isoxazole-5-carbaldehyde
3-Methyl-isoxazole-5-carbaldehyde
Formel: C₅H₅NO₂
Smeltepunkt: 42…44 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09751673
CAS-nummer: 70753-36-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR