4-Methoxypyrimidine-5-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR345706-1G
OR345706-250MG
APOSOR345706-1GEA
1200
DKK
APOSOR345706-1G
APOSOR345706-250MG
4-Methoxypyrimidine-5-carboxylic acid
4-Methoxypyrimidine-5-carboxylic acid
Formel:
C₆H₆N₂O₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD20644991 CAS-nummer: 72411-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...