4-Bromo-3-nitropyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR345055-1G OR345055-250MG
APOSOR345055-1GEA 2780 DKK
APOSOR345055-1G APOSOR345055-250MG
4-Bromo-3-nitropyridine
4-Bromo-3-nitropyridine
Formel: C₅H₃BrN₂O₂
Molekylvægt: 202.994 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04112576
CAS-nummer: 23056-44-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR