N-Methyl-3-(piperidin-4-yl)benzamide hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR321412-1G OR321412-250MG
APOSOR321412-1GEA 5510 DKK
APOSOR321412-1G APOSOR321412-250MG
N-Methyl-3-(piperidin-4-yl)benzamide hydrochloride
N-Methyl-3-(piperidin-4-yl)benzamide hydrochloride
Formel: C₁₃H₁₈N₂O·2ClH
Opbevaringstemperatur: Køleskab
MDL Number: MFCD29056884
CAS-nummer: 1221279-03-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR