D-Norleucinol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (R)-(-)-2-Amino-1-hexanol, (R)-2-Aminohexan-1-ol

OR316035-1G OR316035-5G
APOSOR316035-1GEA 907 DKK
APOSOR316035-1G APOSOR316035-5G
D-Norleucinol 97%
D-Norleucinol
Formel: C₆H₁₅NO
Kogepunkt: 216…218 °C
Smeltepunkt: 40…44 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02683227
CAS-nummer: 80696-28-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR